Ethephon(2-chloroethylphosphonic acid) 90% tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
30163-0010
30163-0050
TS30163-0010EA
212
USD
TS30163-0010
TS30163-0050
Ethephon(2-chloroethylphosphonic acid) 90% tech.
Ethephon(2-chloroethylphosphonic acid)
Formula:
C₂H₆ClO₃P Melting Pt: 74…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002142 CAS Number: 16672-87-0 UN: 2928 ADR: 6.1,II |
Specification Test Results
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 65 to 75 °C |
Titration with NaOH | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...