(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 99%
Supplier: Thermo Scientific Chemicals
33763-0010
33763-0050
33763-0250
TS33763-0010EA
41
USD
TS33763-0010
TS33763-0050
TS33763-0250
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 99%
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Formula:
C₁₀H₁₁NO₂ Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010845 CAS Number: 77943-39-6 |
Specification Test Results
Appearance | White Powder or crystals or flakes |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Specific optical rotation | +165° to +175° (20°C, 589 nm) (c=2, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...