2-Aminoethyl dihydrogen phosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

36742-0050 36742-0250 36742-1000
TS36742-0050EA 31 USD
TS36742-0050 TS36742-0250 TS36742-1000
2-Aminoethyl dihydrogen phosphate 98%
2-Aminoethyl dihydrogen phosphate
Formula: C₂H₈NO₄P
Melting Pt: 241…243 °C
Storage Temperature: Freezer
MDL Number: MFCD00008178
CAS Number: 1071-23-4
EINECS: 213-988-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR