Diisopropyl methylphosphonate 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44938-0050
44938-0250
44938-1000
TS44938-0050EA
41
USD
TS44938-0050
TS44938-0250
TS44938-1000
Diisopropyl methylphosphonate 90%
Diisopropyl methylphosphonate
Formula:
C₇H₁₇O₃P MW: 180.18 g/mol Boiling Pt: 50…51 °C (1 mmHg) Density: 0.985 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00015022 CAS Number: 1445-75-6 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4070 to 1.4130 (20°C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...