3-Quinolineboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Quinoline-3-boronic acid, Quinolin-3-ylboronic acid
36773-0010
36773-0050
36773-0250
TS36773-0010EA
91
USD
TS36773-0010
TS36773-0050
TS36773-0250
3-Quinolineboronic acid 97%
3-Quinolineboronic acid
Formula:
C₉H₈BNO₂ MW: 172.98 g/mol Melting Pt: 148…155 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02183527 CAS Number: 191162-39-7 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Light pink to orange to light brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...