1-Naphthyl phosphate monosodium salt monohydrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate

22689-0010 22689-0050 22689-0250 22689-1000
TS22689-0010EA 36 USD
TS22689-0010 TS22689-0050 TS22689-0250 TS22689-1000
1-Naphthyl phosphate monosodium salt monohydrate 98+%
1-Naphthyl phosphate monosodium salt monohydrate
Formula: C₁₀H₈NaO₄P·1H₂O
MW: 264.15 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00150615
CAS Number: 81012-89-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless to light yellow
Titration with HClO4 ≥98.0 %
Impurity ≤100 ppm Free 1-Naphthol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR