(+)-Dibenzoyl-D-tartaric acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

30251-0250 30251-1000 30251-5000
TS30251-0250EA 56 USD
TS30251-0250 TS30251-1000 TS30251-5000
(+)-Dibenzoyl-D-tartaric acid 98+%
(+)-Dibenzoyl-D-tartaric acid
Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
HPLC ≥98.0 %
Specific optical rotation +117° to +123° (20 °C, 589 nm) (c=5, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR