2-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

43391-0010 43391-0050
TS43391-0010EA 44 USD
TS43391-0010 TS43391-0050
2-(Methoxycarbonyl)phenylboronic acid 97%
2-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
Melting Pt: 101.1…101.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02179452
CAS Number: 374538-03-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR