Methyl (methoxymethylene)acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-methoxyacrylate

33046-0250 33046-1000
TS33046-0250EA 37 USD
TS33046-0250 TS33046-1000 TS33046-5000
Methyl (methoxymethylene)acetate 95%
Methyl (methoxymethylene)acetate
Formula: C₅H₈O₃
MW: 116.1152 g/mol
Boiling Pt: 169 °C (760 mmHg)
Melting Pt: 3…4 °C
Density: 1.07 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00128206
CAS Number: 34846-90-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR