Ethyl (ethoxymethylene)cyanoacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

11848-0010 11848-1000 11848-5000
TS11848-0010EA 294 USD
TS11848-0010 TS11848-1000 TS11848-5000
Ethyl (ethoxymethylene)cyanoacetate 98%
Ethyl (ethoxymethylene)cyanoacetate
Formula: C₈H₁₁NO₃
Boiling Pt: 190…191 °C (30 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00009136
CAS Number: 94-05-3
EINECS: 202-299-5

Order Now

Specification Test Results

Appearance White to yellow to yellow-brown Powder or crystals or flakes or crystalline solid
Infrared spectrum Conforms
Melting point 47 to 53 °C
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Sulfur compounds (as S) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR