Ethyl-3-hydroxybutyrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate

11854-0250 11854-1000 11854-5000
TS11854-0250EA 30 USD
TS11854-0250 TS11854-1000 TS11854-5000
Ethyl-3-hydroxybutyrate 99%
Ethyl-3-hydroxybutyrate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 170 °C (760 mmHg)
Density: 1.01 g/cm³
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00004545
CAS Number: 5405-41-4
EINECS: 226-456-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4195 to 1.4215 (20°C, 589 nm)
GC ≥98.5 %
Ammonium (NH4) ≤50 ppm
Calcium (Ca) ≤600 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤500 ppm
Oxychloride passes test
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR