(S)-(-)-4-Isopropyl-2-oxazolidinone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one

29786-0010 29786-0050 29786-0250
TS29786-0010EA 33 USD
TS29786-0010 TS29786-0050 TS29786-0250
(S)-(-)-4-Isopropyl-2-oxazolidinone 98%
(S)-(-)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00010847
CAS Number: 17016-83-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation -16° to -20° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR