2(5H)-Furanone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Furan-2(5H)-one

37653-0010 37653-0050
TS37653-0010EA 77 USD
TS37653-0010 TS37653-0050
2(5H)-Furanone 95%
2(5H)-Furanone
Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 212…214 °C (760 mmHg)
Melting Pt: 3…4 °C
Density: 1.19 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00005376
CAS Number: 497-23-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to amber Liquid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR