Valeramide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pentanamide

42244-0025 42244-0050 42244-0250
TS42244-0025EA 44 USD
TS42244-0025 TS42244-0050 TS42244-0250
Valeramide 97%
Valeramide
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00041895
CAS Number: 626-97-1
EINECS: 210-974-0

Order Now

Specification Test Results

Appearance White Glistening powder or flakes
Infrared spectrum Conforms
Melting point 101 to 106 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR