Sodium mercaptoacetate (sodium thioglycolate) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt

14889-0010 14889-0500 14889-2500
TS14889-0010EA 393 USD
TS14889-0010 TS14889-0500 TS14889-2500
Sodium mercaptoacetate (sodium thioglycolate) 98%
Sodium mercaptoacetate (sodium thioglycolate)
Formula: HSCH₂COONa
MW: 114.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043386
CAS Number: 367-51-1
EINECS: 206-696-4
UN: 2923
ADR: 8 III

Order Now

Specification Test Results

Appearance White to off-white to light pink Powder
Infrared spectrum Conforms
Titration Iodimetric ≥97.5 %
Elemental Analysis 11.41 to 11.99 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR