Ethyl mercaptoacetate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 2-mercaptoacetate, Ethyl thioglycolate
11867-0025
11867-1000
11867-5000
TS11867-0025EA
529
USD
TS11867-0025
TS11867-1000
TS11867-5000
Ethyl mercaptoacetate 99%
Ethyl mercaptoacetate
Formula:
C₄H₈O₂S MW: 120.17 g/mol Boiling Pt: 54 °C (12 mmHg) Melting Pt: –80 °C Density: 1.09 g/cm³ Flash Pt: 47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004874 CAS Number: 623-51-8 EINECS: 210-800-3 UN: 1228 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4560 to 1.4590 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...