Methyl carbazate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate

16614-0025 16614-1000 16614-5000
TS16614-0025EA 635 USD
TS16614-0025 TS16614-1000 TS16614-5000
Methyl carbazate 97%
Methyl carbazate
Formula: C₂H₆N₂O₂
MW: 90.08 g/mol
Boiling Pt: 108 °C (12 mmHg)
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007594
CAS Number: 6294-89-9
EINECS: 228-560-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to pink Crystalline powder or crystals or platelets
Infrared spectrum Conforms
Melting point 68 to 73 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR