tert-Butyl carbazate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl hydrazinecarboxylate

10788-0250 10788-1000 10788-5000
TS10788-0250EA 90 USD
TS10788-0250 TS10788-1000 TS10788-5000
tert-Butyl carbazate 99%
tert-Butyl carbazate
Formula: C₅H₁₂N₂O₂
MW: 132.16 g/mol
Boiling Pt: 63…65 °C (.1 mmHg)
Melting Pt: 37…41 °C
Density: 1.02 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007593
CAS Number: 870-46-2
EINECS: 212-795-3
UN: 2926
ADR: 4.1 II

Order Now

Specification Test Results

Appearance White to light yellow Lumps
Infrared spectrum Conforms
Melting point 37 to 41 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR