(±)-Ethyl-2-bromopropionate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate

11814-0010 11814-1000 11814-2500
TS11814-0010EA 337 USD
TS11814-0010 TS11814-1000 TS11814-2500
(±)-Ethyl-2-bromopropionate 99%
(±)-Ethyl-2-bromopropionate
Formula: C₅H₉BrO₂
Boiling Pt: 156…160 °C (760 mmHg)
Density: 1.394 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥98.5 %
Sulfate (SO4) ≤300 ppm
Sodium fluosilicate ≤0.1 % (Na2SiF6)
Sulfite (SO2) ≤50 ppm
Iron (Fe) ≤30 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Pb) ≤30 ppm
Titrable acid ≤0.03 meq/g
Titrable base ≤0.01 meq/g
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR