5,6-Dihydro-2H-pyran-2-one 90%

Supplier: Thermo Scientific Chemicals
Warning

42664-0010 42664-0050 42664-0250
TS42664-0010EA 63 USD
TS42664-0010 TS42664-0050 TS42664-0250
5,6-Dihydro-2H-pyran-2-one 90%
5,6-Dihydro-2H-pyran-2-one
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 103 °C (10 mmHg)
Density: 1.139 g/cm³
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010439
CAS Number: 3393-45-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR