2-Hydroxypropyl methacrylate (mixture of isomers) 97+% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester

41215-0010 41215-2500
TS41215-0010EA 103 USD
TS41215-0010 TS41215-2500
2-Hydroxypropyl methacrylate (mixture of isomers) 97+% stabilised
2-Hydroxypropyl methacrylate (mixture of isomers)
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004536
CAS Number: 27813-02-1

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4490 (20°C, 589 nm)
GC ≥97.0 % (Mixture of isomers)
Color scale ≤20 APHA
Stabilizer 150 to 300 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR