Phenylphosphonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylphosphonic Acid

13079-0025 13079-1000 13079-5000
TS13079-0025EA 608 USD
TS13079-0025 TS13079-1000 TS13079-5000
Phenylphosphonic acid 98%
Phenylphosphonic acid
Formula: C₆H₇O₃P
MW: 158.09 g/mol
Melting Pt: 158…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002136
CAS Number: 1571-33-1
EINECS: 216-388-1
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 165 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR