5,5-Dimethylhydantoin 97%

Supplier: Thermo Scientific Chemicals

11631-0010 11631-0051 11631-2500
TS11631-0010EA 86 USD
TS11631-0010 TS11631-0051 TS11631-2500
5,5-Dimethylhydantoin 97%
5,5-Dimethylhydantoin
Formula: C₅H₈N₂O₂
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00005266
CAS Number: 77-71-4
EINECS: 201-051-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 175 to 180 °C
Loss on drying ≤0.25 % (60 °C, 2 h, 1 mmHg)
Titration with NaOH ≥96.0 %
Sulfate (SO4) ≤0.02 %
AsO4, PO4, SiO4 (as SiO2) ≤0.001 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR