5-Chloro-3-pyridineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid

44536-0010 44536-0050
TS44536-0010EA 375 USD
TS44536-0010 TS44536-0050
5-Chloro-3-pyridineboronic acid 95%
5-Chloro-3-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR