4-Ethynyltoluene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethynyl-4-methylbenzene, p-Tolylacetylene
39256-0010
39256-0050
39256-0250
TS39256-0010EA
34
USD
TS39256-0010
TS39256-0050
TS39256-0250
4-Ethynyltoluene 97%
4-Ethynyltoluene
Formula:
C₉H₈ MW: 116.16 g/mol Boiling Pt: 168…170 °C (760 mmHg) Density: 0.916 g/cm³ Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008571 CAS Number: 766-97-2 UN: 3295 ADR: 3 III |
Specification Test Results
Appearance | Light yellow to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5460 to 1.5480 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...