Lithium trifluoromethanesulphonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35871-0050
35871-0250
35871-1000
TS35871-0050EA
32
USD
TS35871-0050
TS35871-0250
TS35871-1000
Lithium trifluoromethanesulphonate 98%
Lithium trifluoromethanesulphonate
Formula:
CF₃LiO₃S Melting Pt: 423 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013227 CAS Number: 33454-82-9 EINECS: 251-528-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Assay | 4.34 to 4.56 % (Li) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...