(±)-Tropic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid
14055-0050
14055-0250
14055-1000
TS14055-0050EA
26
USD
TS14055-0050
TS14055-0250
TS14055-1000
(±)-Tropic acid 97%
DL-Tropic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004255 CAS Number: 529-64-6 EINECS: 208-465-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 115 to 120 °C |
HPLC | ≥96.0 % |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...