Methyl-3-bromopropionate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-bromopropanoate

22179-0250 22179-0500 22179-1000
TS22179-0250EA 87 USD
TS22179-0250 TS22179-0500 TS22179-1000
Methyl-3-bromopropionate 97%
Methyl-3-bromopropionate
Formula: C₄H₇BrO₂
Boiling Pt: 64…66 °C (18 mmHg)
Density: 1.53 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00000250
CAS Number: 3395-91-3
EINECS: 222-247-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4565 to 1.4595 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR