3-Aminobenzene sulphonic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Aminobenzene-3-sulphonic acid, Metanilic acid, 3-Aminobenzenesulphonic acid

41371-0010 41371-1000 41371-2500
TS41371-0010EA 184 USD
TS41371-0010 TS41371-1000 TS41371-2500
3-Aminobenzene sulphonic acid 98%
3-Aminobenzene sulphonic acid
Formula: C₆H₇NO₃S
MW: 173.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00065345
CAS Number: 121-47-1
EINECS: 204-473-6

Order Now

Specification Test Results

Appearance White to grey-beige Powder
Infrared spectrum Conforms
Additional info product may have a purple-pink cast
Titration with NaOH ≥97.5 %
Stabilizer About 0.05% 3,5-di-tert-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR