Diethyl bromomalonate 90+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl 2-bromomalonate

11419-0250 11419-1000 11419-5000
TS11419-0250EA 57 USD
TS11419-0250 TS11419-1000 TS11419-5000
Diethyl bromomalonate 90+%
Diethyl bromomalonate
Formula: C₇H₁₁BrO₄
MW: 239.07 g/mol
Boiling Pt: 233…235 °C (760 mmHg)
Density: 1.4 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009138
CAS Number: 685-87-0
EINECS: 211-683-1
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4560 (20°C, 589 nm)
GC ≥90.0 %
UV at 214 nm A: ≤0.05
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR