5-Bromooxindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole

39853-0010 39853-0050
TS39853-0010EA 38 USD
TS39853-0010 TS39853-0050
5-Bromooxindole 97%
5-Bromooxindole
Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00456998
CAS Number: 20870-78-4

Order Now

Specification Test Results

Appearance Off-white to tan Powder or crystals
Infrared spectrum Conforms
Melting point 220 to 224 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR