(±)-Methyl-2-methylbutyrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl2-methylbutyrate

45023-0010 45023-1000
TS45023-0010EA 142 USD
TS45023-0010 TS45023-0050 TS45023-1000
(±)-Methyl-2-methylbutyrate 98%
(±)-Methyl-2-methylbutyrate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 115 °C (760 mmHg)
Density: 0.885 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00009335
CAS Number: 868-57-5
UN: 3273
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3930 to 1.3960 (20°C, 589 nm)
GC ≥97.5 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR