Fluoranthene 98%

Supplier: Thermo Scientific Chemicals
Warning

11917-0250 11917-1000 11917-5000
TS11917-0250EA 58 USD
TS11917-0250 TS11917-1000 TS11917-5000
Fluoranthene 98%
Fluoranthene
Formula: C₁₆H₁₀
Boiling Pt: 384 °C (760 mmHg)
Melting Pt: 109…111 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001184
CAS Number: 206-44-0
EINECS: 205-912-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Yellow to yellow-green to beige to light grey Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 106 to 111 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR