Benzyl cinnamate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester

21529-0050 21529-1000 21529-5000
TS21529-0050EA 29 USD
TS21529-0050 TS21529-1000 TS21529-5000
Benzyl cinnamate 99%
Benzyl cinnamate
Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Boiling Pt: 195…200 °C (5 mmHg)
Melting Pt: 33…37 °C
Flash Pt: >112 °C
MDL Number: MFCD00004789
CAS Number: 103-41-3
EINECS: 203-109-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Crystalline mass or liquid after melting
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR