3-Phenyl-1-propyne 97% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
37680-0010
37680-0050
37680-0250
TS37680-0010EA
47
USD
TS37680-0010
TS37680-0050
TS37680-0250
3-Phenyl-1-propyne 97% stabilized
3-Phenyl-1-propyne
Formula:
C₉H₈ Boiling Pt: 75 °C (20 mmHg) Density: 0.934 g/cm³ Flash Pt: 52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134431 CAS Number: 10147-11-2 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5260 to 1.5300 (20°C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | 100 to 400 ppm BHT |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...