[2.2]-Paracyclophane 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
29160-0010
29160-0050
29160-0250
TS29160-0010EA
33
USD
TS29160-0010
TS29160-0050
TS29160-0250
[2.2]-Paracyclophane 99%
[2.2]-Paracyclophane
Formula:
C₁₆H₁₆ MW: 208.3 g/mol Melting Pt: 285…288 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003707 CAS Number: 1633-22-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 285 to 288 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...