Bis(4-nitrophenyl)carbonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(4-nitrophenyl) carbonate

17160-0010 17160-0050 17160-0250
TS17160-0010EA 27 USD
TS17160-0010 TS17160-0050 TS17160-0250
Bis(4-nitrophenyl)carbonate 97%
Bis(4-nitrophenyl)carbonate
Formula: C₁₃H₈N₂O₇
MW: 304.22 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00007322
CAS Number: 5070-13-3

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 136 to 144 °C
GC ≥96.0 %
Moisture 6 % to 10 %
Bulk density 475 to 625 kg/m³
Acidity ≤0.1 mg KOH/g (Free Acidity)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR