Methyl methoxyacetate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methoxyacetic methyl ester

16509-0250 16509-1000 16509-5000
TS16509-0250EA 31 USD
TS16509-0250 TS16509-1000 TS16509-5000
Methyl methoxyacetate 99%
Methyl methoxyacetate
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 129…130 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00008451
CAS Number: 6290-49-9
EINECS: 228-539-9
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.3954 to 1.3974 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR