Ethyl acrylate 99.5% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Acrylic acid-ethyl ester

11798-0010 11798-0025 11798-0100 11798-0250
TS11798-0010EA 74 USD
TS11798-0010 TS11798-0025 TS11798-0100 TS11798-0250
Ethyl acrylate 99.5% stabilized
Ethyl acrylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 99 °C (760 mmHg)
Melting Pt: –71 °C
Density: 0.92 g/cm³
Flash Pt: 16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009188
CAS Number: 140-88-5
EINECS: 205-438-8
UN: 1917
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
Water ≤0.05 % (K.F.)
GC ≥99.40 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Free acid ≤0.009 % (as acrylic acid)
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR