3,4-Dihydroxy-3-cyclobutene-1,2-dione 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

15402-0050 15402-0250 15402-1000
TS15402-0050EA 56 USD
TS15402-0050 TS15402-0250 TS15402-1000
3,4-Dihydroxy-3-cyclobutene-1,2-dione 99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: 293 °C
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR