Sodium cyclamate 99%

Supplier: Thermo Scientific Chemicals
Warning

38928-0010 38928-0500 38928-2500
TS38928-0010EA 164 USD
TS38928-0010 TS38928-0500 TS38928-2500
Sodium cyclamate 99%
Sodium cyclamate
Formula: C₆H₁₂NNaO₃S
MW: 201.22 g/mol
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00003827
CAS Number: 139-05-9

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 5.5 to 7.5 (10 % solution)
Titration with HClO4 ≥98.5 % (On dry substance)
Sulfate (SO4) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR