o,o,o-Tritolyl phosphate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36646-0250
36646-1000
36646-5000
TS36646-0250EA
65
USD
TS36646-0250
TS36646-1000
TS36646-5000
o,o,o-Tritolyl phosphate 96%
o,o,o-Tritolyl phosphate
Formula:
C₂₁H₂₁O₄P Boiling Pt: 410 °C (760 mmHg) Melting Pt: –25 °C Density: 1.2 g/cm³ Flash Pt: 225 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041906 CAS Number: 78-30-8 EINECS: 201-103-5 UN: 2574 ADR: 6.1,II Merck Index: 12,09893 |
Specification Test Results
Appearance | Clear colorless to yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5580 to 1.5610 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...