3-Pyridineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid

35965-0010 35965-0050 35965-2500
TS35965-0010EA 74 USD
TS35965-0010 TS35965-0050 TS35965-2500
3-Pyridineboronic acid 95%
3-Pyridineboronic acid
Formula: C₅H₆BNO₂
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674177
CAS Number: 1692-25-7

Order Now

Specification Test Results

Appearance White to yellow to light orange Powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR