(±)-3-Hydroxybutyric acid 95%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
12105-0050
12105-0100
12105-0500
TS12105-0050EA
165
USD
TS12105-0050
TS12105-0100
TS12105-0500
(±)-3-Hydroxybutyric acid 95%, Technical Grade
(±)-3-Hydroxybutyric acid
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 269 Flash Pt: 112 °C Storage Temperature: Freezer |
MDL Number:
MFCD00004546 CAS Number: 300-85-6 EINECS: 206-099-9 Merck Index: 12,04861 |
Specification Test Results
Appearance | Colorless to white to light yellow Solid or liquid |
Infrared spectrum | Conforms |
Titration with NaOH | 94 to 106 % |
HPLC | ≥98.5 % |
Specific optical rotation | +80° to +86° (20°C, 589 nm) (c=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...