4-Tolylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
34443-0010
34443-0050
34443-0250
TS34443-0010EA
41
USD
TS34443-0010
TS34443-0050
TS34443-0250
4-Tolylboronic acid 97%
4-Tolylboronic acid
Formula:
C₇H₉BO₂ Melting Pt: 256…263 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039138 CAS Number: 5720-05-8 |
Specification Test Results
Appearance | White to cream to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 256 to 263 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...