2,2'-Thiodiacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiodiglycolic acid, Thiodiacetic Acid

13874-0010 13874-0250 13874-1000
TS13874-0010EA 215 USD
TS13874-0010 TS13874-0250 TS13874-1000
2,2'-Thiodiacetic acid 98%
2,2'-Thiodiacetic acid
Formula: C₄H₆O₄S
Boiling Pt: 168 °C (14 mmHg)
Melting Pt: 128…131 °C
MDL Number: MFCD00004359
CAS Number: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8,III
Merck Index: 12,09467

Order Now

Specification Test Results

Appearance White to grey Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 131 °C
Titration with NaOH ≥97.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation -36.6° to -40.6° (20 °C, 589 nm) (c=1,4 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR