Methyl crotonate 96%

Supplier: Thermo Scientific Chemicals
Danger

16084-0050 16084-1000 16084-5000
TS16084-0050EA 30 USD
TS16084-0050 TS16084-1000 TS16084-5000
Methyl crotonate 96%
Methyl crotonate
Formula: C₅H₈O₂
Boiling Pt: 118 °C (1013 hPa)
Density: 0.94 g/cm³
Flash Pt: –1 °C
MDL Number: MFCD00009287
CAS Number: 623-43-8
EINECS: 210-793-7
UN: 3272
ADR: 3,II
Merck Index: 13,02625

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4270 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR