Methyl propiolate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester

17404-0100 17404-0500 17404-2500
TS17404-0100EA 109 USD
TS17404-0100 TS17404-0500 TS17404-2500
Methyl propiolate 99%
Methyl propiolate
Formula: C₄H₄O₂
Boiling Pt: 105 °C (1013 hPa)
Density: 0.94 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008572
CAS Number: 922-67-8
EINECS: 213-083-5
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20°C, 589 nm)
GC ≥98.5 %
Chloride (Cl) 8.6 to 9.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR