Methyl chloroacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

14863-0010 14863-0025 14863-0100
TS14863-0010EA 72 USD
TS14863-0010 TS14863-0025 TS14863-0100
Methyl chloroacetate 98%
Methyl chloroacetate
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 130…132 °C (760 mmHg)
Melting Pt: –33 °C
Density: 1.23 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00000931
CAS Number: 96-34-4
EINECS: 202-501-1
UN: 2295
ADR: 6.1,
Merck Index: 12,06122

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4205 to 1.4225 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR