Ethyl chloroacetate 99%

Supplier: Thermo Scientific Chemicals
Danger

11822-0010 11822-0025 11822-0100 11822-2500
TS11822-0010EA 96 USD
TS11822-0010 TS11822-0025 TS11822-0100 TS11822-2500
Ethyl chloroacetate 99%
Ethyl chloroacetate
Formula: C₄H₇ClO₂
Boiling Pt: 142…145 °C (760 mmHg)
Melting Pt: –26 °C
Density: 1.15 g/cm³
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000932
CAS Number: 105-39-5
EINECS: 203-294-0
UN: 1181
ADR: 6.1,II
Merck Index: 13,03818

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR